missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4,5-dimethoxybenzaldehyde, 97%, Thermo Scientific™
£32.80 - £103.40
Chemical Identifiers
CAS | 6948-30-7 |
---|---|
Molecular Formula | C9H9BrO3 |
Molecular Weight (g/mol) | 245.07 |
InChI Key | ICVODPFGWCUVJC-UHFFFAOYSA-N |
PubChem CID | 81379 |
IUPAC Name | 3-bromo-4,5-dimethoxybenzaldehyde |
SMILES | COC1=C(C(=CC(=C1)C=O)Br)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15264794
|
Thermo Scientific Acros
450090010 |
1 g |
£32.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274794
|
Thermo Scientific Acros
450090050 |
5 g | |||||||
Description
Chemical Identifiers
6948-30-7 | |
245.07 | |
81379 | |
COC1=C(C(=CC(=C1)C=O)Br)OC |
C9H9BrO3 | |
ICVODPFGWCUVJC-UHFFFAOYSA-N | |
3-bromo-4,5-dimethoxybenzaldehyde |
Specifications
6948-30-7 | |
96% min. (GC) | |
C9H9BrO3 | |
Solubility in water: insoluble | |
COC1=C(C(=CC(=C1)C=O)Br)OC | |
245.07 | |
245.07 | |
3-Bromo-4, 5-dimethoxybenzaldehyde |
61°C to 63°C | |
Glass bottle | |
5-bromoveratraldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 3,4-dimethoxy-5-bromobenzaldehyde, benzaldehyde,3-bromo-4,5-dimethoxy, benzaldehyde, 3-bromo-4,5-dimethoxy, 5-bromoveratric aldehyde, acmc-209ml7, cambridge id 7187634, 5-bromveratrumaldehyde | |
ICVODPFGWCUVJC-UHFFFAOYSA-N | |
3-bromo-4,5-dimethoxybenzaldehyde | |
81379 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 230-113-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Bromo-4,5-dimethoxybenzaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only