Learn More
3-Bromo-4-cyanopyridine, 95%, Thermo Scientific™
£120.00 - £263.00
Chemical Identifiers
CAS | 13958-98-0 |
---|---|
Molecular Formula | C6H3BrN2 |
Molecular Weight (g/mol) | 183.008 |
MDL Number | MFCD00234143 |
InChI Key | VPDIXJCBBUZNPO-UHFFFAOYSA-N |
Synonym | 3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo |
PubChem CID | 11074051 |
IUPAC Name | 3-bromopyridine-4-carbonitrile |
SMILES | C1=CN=CC(=C1C#N)Br |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13958-98-0 | |
183.008 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
11074051 | |
C1=CN=CC(=C1C#N)Br |
C6H3BrN2 | |
MFCD00234143 | |
3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo | |
3-bromopyridine-4-carbonitrile |
Specifications
13958-98-0 | |
Odorless | |
MFCD00234143 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
3-bromopyridine-4-carbonitrile | |
11074051 | |
95% |
92°C to 94°C | |
C6H3BrN2 | |
3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo | |
C1=CN=CC(=C1C#N)Br | |
183.008 | |
183.01 | |
3-Bromo-4-cyanopyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures