missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 11491878
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160434-49-1
Molecular Formula C9H5BrFO2
Molecular Weight (g/mol) 244.04
MDL Number MFCD00143095
InChI Key ZNIGVADAKXOMQH-DUXPYHPUSA-M
Synonym 3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391
PubChem CID 688312
IUPAC Name (E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid
SMILES [O-]C(=O)\C=C\C1=CC=C(F)C(Br)=C1

Specifications

Melting Point 193°C to 195°C
Quantity 5 g
Beilstein 8977970
Formula Weight 245.05
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorocinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.