missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-heptanone, 98%, Thermo Scientific Chemicals

£46.80 - £250.00

Chemical Identifiers

CAS 42330-10-9
Molecular Formula C7H13BrO
Molecular Weight (g/mol) 193.08
MDL Number MFCD09757526
InChI Key SFKVBRLKXVRUQW-UHFFFAOYNA-N
Synonym 3-bromo-4-heptanone, 4-heptanone, 3-bromo
PubChem CID 11469593
IUPAC Name 3-bromoheptan-4-one
SMILES CCCC(=O)C(Br)CC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11300046
View Documents
Thermo Scientific Alfa Aesar
H32094.03
1 g
£46.80
1g
Please to purchase this item. Need a web account? Register with us today!
11390036
View Documents
Thermo Scientific Alfa Aesar
H32094.09
10 g
£250.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

42330-10-9
193.08
SFKVBRLKXVRUQW-UHFFFAOYNA-N
11469593
CCCC(=O)C(Br)CC
C7H13BrO
MFCD09757526
3-bromo-4-heptanone, 4-heptanone, 3-bromo
3-bromoheptan-4-one

Specifications

42330-10-9
39°C (1 mmHg)
C7H13BrO
MFCD09757526
3-bromo-4-heptanone, 4-heptanone, 3-bromo
CCCC(=O)C(Br)CC
193.08
193.08
3-Bromo-4-heptanone
1.248
75°C (167°F)
1.458
UN3265
SFKVBRLKXVRUQW-UHFFFAOYNA-N
3-bromoheptan-4-one
11469593
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only