missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-4-methylbenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11480403

3-Bromo-4-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11480403
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11480403

Brand: Thermo Scientific Alfa Aesar A11496.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-26-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00002488
InChI Key ZFJOMUKPDWNRFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
PubChem CID 82130
IUPAC Name 3-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1Br)C(O)=O

Specifications

Melting Point 206°C to 211°C
Odor Odorless
Quantity 5 g
Beilstein 1936510
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methylbenzoic acid
compliance-icons
Product Identifier
  • 3-Bromo-4-methylbenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-Bromo-4-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-4-methylbenzoic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.