missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(hydroxymethyl)pyridine, 97%, Thermo Scientific™

£377.41 - £1293.53

Chemical Identifiers

CAS 37669-64-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD03265757
InChI Key WDVDHJLKXYCOFS-UHFFFAOYSA-N
Synonym 5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
PubChem CID 2784733
IUPAC Name (5-bromopyridin-3-yl)methanol
SMILES OCC1=CC(Br)=CN=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11906461
View Documents Promotion Details
Thermo Scientific Acros
444050010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11926461
View Documents Promotion Details
Thermo Scientific Acros
444050050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

37669-64-0
188.02
WDVDHJLKXYCOFS-UHFFFAOYSA-N
2784733
OCC1=CC(Br)=CN=C1
C6H6BrNO
MFCD03265757
5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
(5-bromopyridin-3-yl)methanol

Specifications

37669-64-0
100.0
Yellow
Authentic
Glass Bottle
MFCD03265757
WDVDHJLKXYCOFS-UHFFFAOYSA-N
(5-bromopyridin-3-yl)methanol
2784733
97%
3-Bromo-5-(hydroxymethyl)pyridine
96.0
27.0°C
154.0°C (4.0 Torr)
96% min. (GC)
C6H6BrNO
5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
OCC1=CC(Br)=CN=C1
188.02
188.02
Liquid or Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 3-Bromo-5-(hydroxymethyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only