missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(pentafluorothio)benzoic acid, 97%, Thermo Scientific™

Product Code. 15487127
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1180675-98-2
Molecular Formula C7H4BrF5O2S
Molecular Weight (g/mol) 327.063
MDL Number MFCD16652482
InChI Key GGDWTFSLWQRHPR-UHFFFAOYSA-N
Synonym 3-bromo-5-pentafluoro-??-sulfanyl benzoic acid, 3-bromo-5-pentafluorothio benzoic acid, 3-bromo-5-pentafluorosulfur benzoic acid, 3-bromo-5-pentafluorosulfanyl benzoic acid, 3-bromo-5-pentafluoro-lambda6-sulphanyl benzoic acid
PubChem CID 58137421
IUPAC Name 3-bromo-5-(pentafluoro-$l^{6}-sulfanyl)benzoic acid
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Br)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 247.16
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(pentafluorothio)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'