missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chlorochromone, 96%, Thermo Scientific™

Product Code. 15474717
Click to view available options
Quantity:
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73220-38-9
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.48
MDL Number MFCD02093123
InChI Key ZILSBPAUJJBEFF-UHFFFAOYSA-N
Synonym 3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
PubChem CID 3498945
IUPAC Name 3-bromo-6-chlorochromen-4-one
SMILES ClC1=CC=C2OC=C(Br)C(=O)C2=C1

Specifications

Melting Point 128°C to 130°C
Quantity 2 g
Formula Weight 259.49
Percent Purity 96%
Chemical Name or Material 3-Bromo-6-chlorochromone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'