missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-7-nitroindole, 97%, Thermo Scientific™

Product Code. 15420929
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397864-11-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
MDL Number MFCD02684155
InChI Key ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
PubChem CID 17750599
IUPAC Name 3-bromo-7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2Br

Specifications

Quantity 1 g
Synonym 3-bromo-7-nitroindole, 1h-indole, 3-bromo-7-nitro, 1h-indole,3-bromo-7-nitro, 6-methoxy-quinolin-3-ylamine
Formula Weight 241.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-7-nitroindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'