missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N-(3-pyridylmethyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15457458
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849136-14-7
Molecular Formula C12H11BrN2O2S
Molecular Weight (g/mol) 327.196
MDL Number MFCD07356489
InChI Key VAQOTDUYHINTAB-UHFFFAOYSA-N
Synonym 3-bromo-n-pyridin-3-ylmethyl benzenesulfonamide, 3-bromo-n-3-pyridylmethyl benzenesulfonamide, 3-bromo-n-pyridin-3-yl methyl benzene-1-sulfonamide
PubChem CID 3772600
IUPAC Name 3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CN=CC=C2

Specifications

Quantity 1 g
Formula Weight 327.2
Percent Purity 97%
Chemical Name or Material 3-Bromo-N-(3-pyridylmethyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'