missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-N,N-dimethylaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.
Solubility
Miscible with chloroform, dichloromethane and methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.
Solubility
Miscible with chloroform, dichloromethane and methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 11°C |
Density | 1.365 |
Boiling Point | 126°C (14 mmHg) |
Refractive Index | 1.6015 |
Quantity | 5 g |
Beilstein | 2079908 |
Solubility Information | Miscible with chloroform,dichloromethane and methanol. |
Formula Weight | 200.08 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-N,N-dimethylaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'