missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzanthrone, tech. 85%

Product Code. p-7044616
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11381907

Brand: Thermo Scientific Alfa Aesar L07422.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81-96-9 | C17H9BrO | 309.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-96-9
Molecular Formula C17H9BrO
Molecular Weight (g/mol) 309.162
MDL Number MFCD00021094
InChI Key WVECFEIAZAKUNF-UHFFFAOYSA-N
Synonym 3-bromobenzanthrone, 3-brombenzanthrone, benzanthrone, 3-bromo, 7-bromomesobenzanthrone, 3-bromo-7h-benzo de anthracen-7-one, 7h-benz de anthracen-7-one, 3-bromo, 3-bromobenz d,e anthrone, 3-bromo-7h-benz de anthracen-7-one, 3-bromo-7h-benz de anthracene-7-one, unii-3dh9916fno
PubChem CID 6696
IUPAC Name 3-bromobenzo[b]phenalen-7-one
SMILES C1=CC=C2C(=C1)C3=C4C(=C(C=C3)Br)C=CC=C4C2=O

Specifications

Melting Point 164°C to 168°C
Quantity 25 g
Beilstein 1975512
Formula Weight 309.17
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Bromobenzanthrone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.