missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11945891
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 34.6°C
CAS 107549-22-4, 60-29-7
Color Yellow
CAS Min % 5.37, 87.47
CAS Max % 12.53, 94.63
Physical Form Liquid
Quantity 100 mL
Molecular Formula C7H6Br2Mg
MDL Number MFCD01319902
Flash Point −40°C
Synonym 3-bromobenzylmagnesium bromide, 3-bromobenzylrnagnesium bromide, 3-bromobenzyl magnesium bromide, ohxweozzkbjutd-uhfffaoysa-m, magnesium,bromo 3-bromophenyl methyl, bromo 3-bromophenyl methyl magnesium, magnesium,1-bromo-3-methanidylbenzene,bromide, 3-bromobenzylmagnesium bromide 0.25 m in diethyl ether, 3-bromobenzylmagnesium bromide, 0.25m in diethyl ether, 3-bromobenzylmagnesium bromide solution, 0.25 m in diethyl ether, grignard reagent
InChI Key OHXWEOZZKBJUTD-UHFFFAOYSA-M
SMILES Br[Mg]CC1=CC(Br)=CC=C1
IUPAC Name magnesium;1-bromo-3-methanidylbenzene;bromide
Molecular Weight (g/mol) 274.24
PubChem CID 16217187
Formula Weight 274.24
Concentration or Composition (by Analyte or Components) 0.15 to 0.35M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.766
Density 0.7660g/mL
Chemical Name or Material 3-Bromobenzylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'