Learn More
3-Bromocyclohexene, 95%, Thermo Scientific Chemicals
£54.10 - £675.00
Chemical Identifiers
CAS | 1521-51-3 |
---|---|
Molecular Formula | C6H9Br |
Molecular Weight (g/mol) | 161.04 |
MDL Number | MFCD00013775 |
InChI Key | AJKDUJRRWLQXHM-UHFFFAOYSA-N |
Synonym | 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene |
PubChem CID | 137057 |
IUPAC Name | 3-bromocyclohexene |
SMILES | C1CC=CC(C1)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326741
|
Thermo Scientific Acros
335120100 |
10 mL | Glass bottle |
£54.10
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10029540
|
Thermo Scientific Acros
335120500 |
50 mL | Glass bottle |
£172.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10627164
|
Thermo Scientific Acros
335122500 |
250 mL | Glass bottle |
£675.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1521-51-3 | |
161.04 | |
AJKDUJRRWLQXHM-UHFFFAOYSA-N | |
137057 | |
C1CC=CC(C1)Br |
C6H9Br | |
MFCD00013775 | |
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene | |
3-bromocyclohexene |
Specifications
1521-51-3 | |
100.0 | |
1.3800g/mL | |
54°C | |
94% min. (GC) | |
1.5280 to 1.5320 | |
1.38 | |
AJKDUJRRWLQXHM-UHFFFAOYSA-N | |
3-bromocyclohexene | |
137057 | |
95% | |
3-Bromocyclohexene |
94.0 | |
White | |
64.0°C to 65.0°C (15.0 mmHg) | |
Authentic | |
C6H9Br | |
MFCD00013775 | |
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene | |
C1CC=CC(C1)Br | |
161.04 | |
161.04 | |
Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
Recommended Storage : May darken on ageing
- 3-Bromocyclohexene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only