missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 98%, Thermo Scientific Chemicals

Product Code. 10474755
Click to view available options
Quantity:
2.5 mL
10 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.6350g/mL
Boiling Point 103.0°C (750.0 mmHg)
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Quantity 10 mL
Specific Gravity 1.635
Stabilizer copper
Formula Weight 146.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromofuran, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromofuran
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.