missing translation for 'onlineSavingsMsg'
Learn More
3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals Product Code.: 15496497

3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals

Product Code. 15496497
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15496497

Brand: Thermo Scientific Alfa Aesar H33652.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92404-63-2
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD19687763
InChI Key KHMAAFDMZSYVEF-UHFFFAOYSA-N
Synonym 3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
PubChem CID 641477
IUPAC Name 3-bromofuro[3,2-b]pyridine
SMILES C1=CC2=C(C(=CO2)Br)N=C1

Specifications

Melting Point 51°C to 53°C
Quantity 5 g
Formula Weight 198.02
Percent Purity 96%
Chemical Name or Material 3-Bromofuro[3,2-b]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10202998 Certificate TypeCertificate of Analysis Date19/05/2023 Catalog NumberH33652.03,H33652.06
Lot Number10197281 Certificate TypeCertificate of Analysis Date19/05/2023 Catalog NumberH33652.03,H33652.06
Lot Number10170078 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33652.03,H33652.06,H33652.MD
Lot Number10197281 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33652.03,H33652.06,H33652.MD
Lot Number10185035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33652.03,H33652.06,H33652.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.