missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-5-chlorobenzo[b]thiophene, 95+%, Thermo Scientific™

Product Code. 10713633
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1198-51-2
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.56
MDL Number MFCD00173769
InChI Key FKQSFVITUNJLCY-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553
PubChem CID 2779857
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
SMILES ClC1=CC=C2SC=C(CBr)C2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 261.57
Percent Purity 95%
Chemical Name or Material 3-(Bromomethyl)-5-chlorobenzo[b]thiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'