missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopropionic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.
Solubility
Soluble in water, alcohol, ether, chloroform, benzene
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.
Solubility
Soluble in water, alcohol, ether, chloroform, benzene
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
Melting Point | 57°C to 63°C |
Density | 1.48 |
Boiling Point | 140°C to 142°C (45 mmHg) |
Flash Point | 65°C (149°F) |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 1071333 |
Merck Index | 14,1433 |
Solubility Information | Soluble in water, alcohol, ether, chloroform, benzene |
Formula Weight | 152.98 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'