missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic acid, 97%

Product Code. 10023900 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 10023900

Brand: Thermo Scientific Acros 325771000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-92-1 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-92-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00002763
InChI Key DHXNZYCXMFBMHE-UHFFFAOYSA-N
Synonym 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
PubChem CID 11553
IUPAC Name 3-bromopropanoic acid
SMILES OC(=O)CCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 63°C
Color White to Yellow
Boiling Point 140°C to 142°C (45.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula BrCH2CH2CO2H
Quantity 100 g
Beilstein 02, 256
Merck Index 15, 1444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chcl3 and benzene
Formula Weight 152.98
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Bromopropionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.