missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-sulfonyl chloride, 90%, Thermo Scientific™

Catalog Number 10527284
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
170727-02-3
C4H2BrClO2S2
261.532
MFCD08059484
OPLMNCCLQDVCTP-UHFFFAOYSA-N
3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo
13675598
3-bromothiophene-2-sulfonyl chloride
C1=CSC(=C1Br)S(=O)(=O)Cl
This item is not returnable. View return policy
170727-02-3
C4H2BrClO2S2
261.532
MFCD08059484
OPLMNCCLQDVCTP-UHFFFAOYSA-N
3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo
13675598
3-bromothiophene-2-sulfonyl chloride
C1=CSC(=C1Br)S(=O)(=O)Cl
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 261.55
Percent Purity 90%
Chemical Name or Material 3-Bromothiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'