missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene, 97%, Thermo Scientific Chemicals

Artikelnummer. 10346980
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1

Spezifikation

Color Colorless to Yellow
Density 1.7400g/mL
Boiling Point 158.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5905 to 1.5925
Quantity 25 mL
Specific Gravity 1.74
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in acetone,benzene,slightly soluble in chloroform
Formula Weight 163.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromothiophene
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • 3-Bromothiophene
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'