Learn More
3-Chloro-1,2-benzisothiazole, 97+%, Thermo Scientific Chemicals
£56.90 - £192.00
Chemical Identifiers
CAS | 7716-66-7 |
---|---|
Molecular Formula | C7H4ClNS |
Molecular Weight (g/mol) | 169.626 |
MDL Number | MFCD00673254 |
InChI Key | BCPVKLRBQLRWDQ-UHFFFAOYSA-N |
Synonym | 3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole |
PubChem CID | 598190 |
IUPAC Name | 3-chloro-1,2-benzothiazole |
SMILES | C1=CC=C2C(=C1)C(=NS2)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394755
|
Thermo Scientific Alfa Aesar
L17791.06 |
5 g |
£56.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304765
|
Thermo Scientific Alfa Aesar
L17791.14 |
25 g |
£192.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.
Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
7716-66-7 | |
169.626 | |
BCPVKLRBQLRWDQ-UHFFFAOYSA-N | |
598190 | |
C1=CC=C2C(=C1)C(=NS2)Cl |
C7H4ClNS | |
MFCD00673254 | |
3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole | |
3-chloro-1,2-benzothiazole |
Specifications
7716-66-7 | |
80°C to 86°C (0.75 mmHg) | |
MFCD00673254 | |
3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole | |
BCPVKLRBQLRWDQ-UHFFFAOYSA-N | |
3-chloro-1,2-benzothiazole | |
598190 | |
99% |
37°C to 44°C | |
C7H4ClNS | |
119370 | |
Soluble in Chloroform,Dichloromethane and Ethyl Acetate. | |
C1=CC=C2C(=C1)C(=NS2)Cl | |
169.626 | |
169.63 | |
3-Chloro-1,2-benzisothiazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only