missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10400774
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 63425-54-7
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.55
MDL Number MFCD09879994
InChI Key GWZMWAZUCSGVLQ-UHFFFAOYSA-N
Synonym 3-chloro-1-methyl-1h-pyrazole, 1h-pyrazole, 3-chloro-1-methyl
PubChem CID 13630086
IUPAC Name 3-chloro-1-methylpyrazole
SMILES CN1C=CC(Cl)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 116.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'