missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzamide, 97+%, Thermo Scientific™

Product Code. 11491177
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104326-94-5
Molecular Formula C7H5ClFNO
Molecular Weight (g/mol) 173.571
MDL Number MFCD00042205
InChI Key KGTCFPJGIYHDRZ-UHFFFAOYSA-N
Synonym benzamide, 3-chloro-2-fluoro, acmc-1c9g7, benzamide,3-chloro-2-fluoro, 3-chloranyl-2-fluoranyl-benzamide
PubChem CID 517842
IUPAC Name 3-chloro-2-fluorobenzamide
SMILES C1=CC(=C(C(=C1)Cl)F)C(=O)N

Specifications

Melting Point 124°C to 127°C
Quantity 10 g
Formula Weight 173.58
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-2-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'