missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzylamine, 97%, Thermo Scientific™

Product Code. 11354096
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72235-55-3
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD01631391
InChI Key LYKWZKBLNOCJMA-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobenzylamine, 3-chloro-2-fluorophenyl methanamine, 3-chloro-2-fluoro-benzylamine, 3-chloro-2-fluorophenyl methylamine, benzenemethanamine, 3-chloro-2-fluoro, 1-3-chloro-2-fluorophenyl methanamine, 2-fluoro-3-chlorobenzylamine, 2-fluoro-3-chlorobenzyl amine, 3-chloro-2-fluoro benzylamine, rarechem al bw 1387
PubChem CID 2773614
IUPAC Name (3-chloro-2-fluorophenyl)methanamine
SMILES C1=CC(=C(C(=C1)Cl)F)CN

Specifications

Refractive Index 1.5405
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'