missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-iodotoluene, 98%, Thermo Scientific™

Product Code. 11366996
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5100-98-1
Molecular Formula C7H6ClI
Molecular Weight (g/mol) 252.479
MDL Number MFCD00051756
InChI Key FTGLKPMFTLNUBN-UHFFFAOYSA-N
Synonym 3-chloro-2-iodotoluene, toluene, 3-chloro-2-iodo, benzene, 1-chloro-2-iodo-3-methyl, pubchem3655, acmc-1asrb, ksc493o0b, 1-chloro-2-iodo-3-methyl-benzene, buttpark 121\11-21, 1-chloranyl-2-iodanyl-3-methyl-benzene
PubChem CID 138414
IUPAC Name 1-chloro-2-iodo-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)I

Specifications

Melting Point 25°C to 29°C
Boiling Point 134°C to 136°C (20 mmHg)
Quantity 1 g
Beilstein 2961541
Sensitivity Light sensitive
Formula Weight 252.48
Percent Purity 98%
Chemical Name or Material 3-Chloro-2-iodotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'