missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-iodotoluene, 98%, Thermo Scientific™
£33.20 - £104.00
Chemical Identifiers
CAS | 5100-98-1 |
---|---|
Molecular Formula | C7H6ClI |
Molecular Weight (g/mol) | 252.479 |
MDL Number | MFCD00051756 |
InChI Key | FTGLKPMFTLNUBN-UHFFFAOYSA-N |
Synonym | 3-chloro-2-iodotoluene, toluene, 3-chloro-2-iodo, benzene, 1-chloro-2-iodo-3-methyl, pubchem3655, acmc-1asrb, ksc493o0b, 1-chloro-2-iodo-3-methyl-benzene, buttpark 121\11-21, 1-chloranyl-2-iodanyl-3-methyl-benzene |
PubChem CID | 138414 |
IUPAC Name | 1-chloro-2-iodo-3-methylbenzene |
SMILES | CC1=C(C(=CC=C1)Cl)I |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5100-98-1 | |
252.479 | |
FTGLKPMFTLNUBN-UHFFFAOYSA-N | |
138414 | |
CC1=C(C(=CC=C1)Cl)I |
C7H6ClI | |
MFCD00051756 | |
3-chloro-2-iodotoluene, toluene, 3-chloro-2-iodo, benzene, 1-chloro-2-iodo-3-methyl, pubchem3655, acmc-1asrb, ksc493o0b, 1-chloro-2-iodo-3-methyl-benzene, buttpark 121\11-21, 1-chloranyl-2-iodanyl-3-methyl-benzene | |
1-chloro-2-iodo-3-methylbenzene |
Specifications
5100-98-1 | |
134°C to 136°C (20 mmHg) | |
MFCD00051756 | |
Light sensitive | |
FTGLKPMFTLNUBN-UHFFFAOYSA-N | |
1-chloro-2-iodo-3-methylbenzene | |
138414 | |
98% |
25°C to 29°C | |
C7H6ClI | |
2961541 | |
3-chloro-2-iodotoluene, toluene, 3-chloro-2-iodo, benzene, 1-chloro-2-iodo-3-methyl, pubchem3655, acmc-1asrb, ksc493o0b, 1-chloro-2-iodo-3-methyl-benzene, buttpark 121\11-21, 1-chloranyl-2-iodanyl-3-methyl-benzene | |
CC1=C(C(=CC=C1)Cl)I | |
252.479 | |
252.48 | |
3-Chloro-2-iodotoluene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures