Learn More
3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™
£21.50 - £57.40
Chemical Identifiers
CAS | 80563-86-6 |
---|---|
Molecular Formula | C7H6Cl2O2S |
Molecular Weight (g/mol) | 225.09 |
MDL Number | MFCD00052198 |
InChI Key | ZSIYKAQPQRTBPF-UHFFFAOYSA-N |
Synonym | 3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride |
PubChem CID | 522734 |
IUPAC Name | 3-chloro-2-methylbenzenesulfonyl chloride |
SMILES | CC1=C(C=CC=C1Cl)S(=O)(=O)Cl |
Description
Chemical Identifiers
80563-86-6 | |
225.09 | |
ZSIYKAQPQRTBPF-UHFFFAOYSA-N | |
522734 | |
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl |
C7H6Cl2O2S | |
MFCD00052198 | |
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride | |
3-chloro-2-methylbenzenesulfonyl chloride |
Specifications
80563-86-6 | |
100.0 | |
White to Yellow | |
97.5% min. (ex Cl) (Argentometry) | |
Cl(CH3)C6H3SO2Cl | |
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride | |
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl | |
225.09 | |
225.09 | |
Crystals |
97.5 | |
71°C to 73°C | |
Authentic | |
C7H6Cl2O2S | |
MFCD00052198 | |
ZSIYKAQPQRTBPF-UHFFFAOYSA-N | |
3-chloro-2-methylbenzenesulfonyl chloride | |
522734 | |
98% | |
3-chloro-2-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 3-Chloro-2-methylbenzenesulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only