Learn More
3-Chloro-2-methylpropene, 90%, tech., Thermo Scientific Chemicals
Chemical Identifiers
CAS | 563-47-3 |
---|---|
Molecular Formula | C4H7Cl |
Molecular Weight (g/mol) | 90.55 |
MDL Number | MFCD00000953 |
InChI Key | OHXAOPZTJOUYKM-UHFFFAOYSA-N |
Synonym | 3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile |
PubChem CID | 11241 |
ChEBI | CHEBI:82419 |
IUPAC Name | 3-chloro-2-methylprop-1-ene |
SMILES | CC(=C)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031580
|
Thermo Scientific Acros
148420500 |
50 mL | Glass bottle |
N/A
|
N/A | ||||
10457920
|
Thermo Scientific Acros
148422500 |
250 mL | Glass bottle |
N/A
|
N/A | ||||
10447920
|
Thermo Scientific Acros
148420010 |
1 L | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
563-47-3 | |
90.55 | |
OHXAOPZTJOUYKM-UHFFFAOYSA-N | |
11241 | |
3-chloro-2-methylprop-1-ene |
C4H7Cl | |
MFCD00000953 | |
3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile | |
CHEBI:82419 | |
CC(=C)CCl |
Specifications
563-47-3 | |
100.0 | |
0.9200g/mL | |
-18°C | |
88% min. (GC) | |
H2C=C(CH3)CH2Cl | |
MFCD00000953 | |
15, 2150 | |
3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile | |
OHXAOPZTJOUYKM-UHFFFAOYSA-N | |
3-chloro-2-methylprop-1-ene | |
11241 | |
0.42 mPa.s (20°C) | |
90% | |
Liquid |
88.0 | |
-80°C | |
71°C to 72°C | |
Authentic | |
C4H7Cl | |
1.4262 to 1.4282 | |
01, 209 | |
0.92 | |
Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CC(=C)CCl | |
90.55 | |
CHEBI:82419 | |
90.55 | |
Technical | |
3-Chloro-2-methylpropene, tech., 90% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapo
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-251-2
missing translation for 'rtecsNumber' : UC8050000
missing translation for 'tsca' : TSCA
- 3-Chloro-2-methylpropene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only