missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
£82.00 - £240.00
Chemical Identifiers
CAS | 850589-50-3 |
---|---|
Molecular Formula | C12H15BClNO3 |
Molecular Weight (g/mol) | 267.516 |
MDL Number | MFCD07363775 |
InChI Key | AQTGPCIHLDJZJF-UHFFFAOYSA-N |
Synonym | 3-chloro-4-piperidine-1-carbonyl phenylboronic acid, 3-chloro-4-piperidine-1-carbonyl phenyl boronic acid, 3-chloro-4-piperidin-1-ylcarbonyl benzeneboronic acid, boronic acid,b-3-chloro-4-1-piperidinylcarbonyl phenyl, 1-4-borono-2-chlorobenzoyl piperidine, n-piperidinyl 2-chloro-4-boronobenzamide, 4-borono-2-chlorophenyl piperidin-1-yl methanone, 3-chloro-4-1-piperidinylcarbonyl benzeneboronic acid, b-3-chloro-4-1-piperidinylcarbonyl phenyl-boronic acid |
PubChem CID | 44119648 |
IUPAC Name | [3-chloro-4-(piperidine-1-carbonyl)phenyl]boronic acid |
SMILES | B(C1=CC(=C(C=C1)C(=O)N2CCCCC2)Cl)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
850589-50-3 | |
267.516 | |
AQTGPCIHLDJZJF-UHFFFAOYSA-N | |
44119648 | |
B(C1=CC(=C(C=C1)C(=O)N2CCCCC2)Cl)(O)O |
C12H15BClNO3 | |
MFCD07363775 | |
3-chloro-4-piperidine-1-carbonyl phenylboronic acid, 3-chloro-4-piperidine-1-carbonyl phenyl boronic acid, 3-chloro-4-piperidin-1-ylcarbonyl benzeneboronic acid, boronic acid,b-3-chloro-4-1-piperidinylcarbonyl phenyl, 1-4-borono-2-chlorobenzoyl piperidine, n-piperidinyl 2-chloro-4-boronobenzamide, 4-borono-2-chlorophenyl piperidin-1-yl methanone, 3-chloro-4-1-piperidinylcarbonyl benzeneboronic acid, b-3-chloro-4-1-piperidinylcarbonyl phenyl-boronic acid | |
[3-chloro-4-(piperidine-1-carbonyl)phenyl]boronic acid |
Specifications
850589-50-3 | |
C12H15BClNO3 | |
3-chloro-4-piperidine-1-carbonyl phenylboronic acid, 3-chloro-4-piperidine-1-carbonyl phenyl boronic acid, 3-chloro-4-piperidin-1-ylcarbonyl benzeneboronic acid, boronic acid,b-3-chloro-4-1-piperidinylcarbonyl phenyl, 1-4-borono-2-chlorobenzoyl piperidine, n-piperidinyl 2-chloro-4-boronobenzamide, 4-borono-2-chlorophenyl piperidin-1-yl methanone, 3-chloro-4-1-piperidinylcarbonyl benzeneboronic acid, b-3-chloro-4-1-piperidinylcarbonyl phenyl-boronic acid | |
B(C1=CC(=C(C=C1)C(=O)N2CCCCC2)Cl)(O)O | |
267.516 | |
267.52 | |
3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid |
160°C to 163°C | |
MFCD07363775 | |
AQTGPCIHLDJZJF-UHFFFAOYSA-N | |
[3-chloro-4-(piperidine-1-carbonyl)phenyl]boronic acid | |
44119648 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures