missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-cyanobenzeneboronic acid, 96%, Thermo Scientific Chemicals
£124.00 - £426.00
Chemical Identifiers
CAS | 1008415-02-8 |
---|---|
Molecular Formula | C7H5BClNO2 |
Molecular Weight (g/mol) | 181.382 |
InChI Key | QCJTUOIMDNJEHR-UHFFFAOYSA-N |
Synonym | 3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid |
PubChem CID | 46738932 |
IUPAC Name | (3-chloro-4-cyanophenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)C#N)Cl)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498098
|
Thermo Scientific Alfa Aesar
H61543.MD |
250 mg |
£124.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488098
|
Thermo Scientific Alfa Aesar
H61543.03 |
1 g |
£426.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1008415-02-8 | |
181.382 | |
3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid | |
(3-chloro-4-cyanophenyl)boronic acid |
C7H5BClNO2 | |
QCJTUOIMDNJEHR-UHFFFAOYSA-N | |
46738932 | |
B(C1=CC(=C(C=C1)C#N)Cl)(O)O |
Specifications
1008415-02-8 | |
3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid | |
B(C1=CC(=C(C=C1)C#N)Cl)(O)O | |
181.382 | |
164.93 | |
Solid |
C7H5BClNO2 | |
QCJTUOIMDNJEHR-UHFFFAOYSA-N | |
(3-chloro-4-cyanophenyl)boronic acid | |
46738932 | |
96% | |
3-Chloro-4-cyanobenzeneboronic acid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only