missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-ethoxy-5-fluorophenol, 97%, Thermo Scientific™

Product Code. 15465397
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-82-3
Molecular Formula C8H8ClFO2
Molecular Weight (g/mol) 190.598
MDL Number MFCD09832339
InChI Key SBGFBMVWWJYFAI-UHFFFAOYSA-N
Synonym 3-chloro-4-ethoxy-5-fluorophenol, jrd
PubChem CID 20111913
IUPAC Name 3-chloro-4-ethoxy-5-fluorophenol
SMILES CCOC1=C(C=C(C=C1Cl)O)F

Specifications

Melting Point 86°C to 90°C
Quantity 1 g
Formula Weight 190.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-ethoxy-5-fluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'