missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-ethoxy-5-fluorophenylacetonitrile, 97%, Thermo Scientific™

Product Code. 15435617
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-94-7
Molecular Formula C10H9ClFNO
Molecular Weight (g/mol) 213.636
MDL Number MFCD09832343
InChI Key SYLGCFDLLYOKTE-UHFFFAOYSA-N
Synonym 3-chloro-4-ethoxy-5-fluorophenylacetonitrile, 2-3-chloro-4-ethoxy-5-fluorophenyl acetonitrile, 3-chloro-4-ethoxy-5-fluorophenylacetonitrile, jrd, 2-3-chloro-4-ethoxy-5-fluorophenyl ethanenitrile
PubChem CID 46737660
IUPAC Name 2-(3-chloro-4-ethoxy-5-fluorophenyl)acetonitrile
SMILES CCOC1=C(C=C(C=C1Cl)CC#N)F

Specifications

Melting Point 32°C to 34°C
Quantity 1 g
UN Number UN3439
Formula Weight 213.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-ethoxy-5-fluorophenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'