missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-ethoxyphenylboronic acid, 98%, Thermo Scientific™
£42.38 - £136.49
Chemical Identifiers
CAS | 279261-81-3 |
---|---|
Molecular Formula | C8H10BClO3 |
Molecular Weight (g/mol) | 200.43 |
InChI Key | QZMQZKMQXURQNC-UHFFFAOYSA-N |
Synonym | 3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride |
PubChem CID | 3717658 |
IUPAC Name | (3-chloro-4-ethoxyphenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)OCC)Cl)(O)O |
Description
Chemical Identifiers
279261-81-3 | |
200.43 | |
3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride | |
(3-chloro-4-ethoxyphenyl)boronic acid |
C8H10BClO3 | |
QZMQZKMQXURQNC-UHFFFAOYSA-N | |
3717658 | |
B(C1=CC(=C(C=C1)OCC)Cl)(O)O |
Specifications
279261-81-3 | |
Authentic | |
Glass Bottle | |
3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride | |
B(C1=CC(=C(C=C1)OCC)Cl)(O)O | |
200.43 | |
200.43 | |
3-Chloro-4-ethoxyphenylboronic acid |
69.0°C to 70.0°C | |
97.5% min. (HPLC) | |
C8H10BClO3 | |
QZMQZKMQXURQNC-UHFFFAOYSA-N | |
(3-chloro-4-ethoxyphenyl)boronic acid | |
3717658 | |
98% |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Chloro-4-ethoxyphenylboronic acid, 98%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only