missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenol, 99%, Thermo Scientific™

£24.90 - £71.16

Chemical Identifiers

CAS 2613-23-2
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00002257
InChI Key ZQXLIXHVJVAPLW-UHFFFAOYSA-N
Synonym phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
PubChem CID 75790
IUPAC Name 3-chloro-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Cl)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10252022
View Documents Promotion Details
Thermo Scientific Acros
277200050
5 g Glass bottle
£24.90
5g
Please to purchase this item. Need a web account? Register with us today!
10491481
View Documents Promotion Details
Thermo Scientific Acros
277200250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

2613-23-2
146.55
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
75790
C1=CC(=C(C=C1O)Cl)F
C6H4ClFO
MFCD00002257
phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
3-chloro-4-fluorophenol

Specifications

2613-23-2
100.0
Beige to Brown
108°C
99%
ClC6H3FOH
phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
3-chloro-4-fluorophenol
75790
99%
3-Chloro-4-fluorophenol, 99%
98.5
39°C to 44°C
104°C (11.0 mmHg)
Authentic
C6H4ClFO
MFCD00002257
Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
C1=CC(=C(C=C1O)Cl)F
146.55
146.55
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 220-042-5

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Chloro-4-fluorophenol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only