missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylboronic acid, 97%, ACROS Organics™

Catalog Number p-2690343
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
144432-85-9
C6H5BClFO2
174.362
MFCD00051800
WJDZZXIDQYKVDG-UHFFFAOYSA-N
3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
2734660
(3-chloro-4-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)F)Cl)(O)O
This item is not returnable. View return policy
144432-85-9
C6H5BClFO2
174.362
MFCD00051800
WJDZZXIDQYKVDG-UHFFFAOYSA-N
3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
2734660
(3-chloro-4-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)F)Cl)(O)O
CAS Min % 96.0
CAS Max % 100.0
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.37
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 3-Chloro-4-fluorophenylboronic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'