Learn More
3-Chloro-4-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
£44.50 - £433.00
Chemical Identifiers
CAS | 37908-96-6 |
---|---|
Molecular Formula | C8H7ClO3 |
Molecular Weight (g/mol) | 186.591 |
MDL Number | MFCD00016512 |
InChI Key | IBCQUQXCTOPJOD-UHFFFAOYSA-N |
Synonym | 3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid |
PubChem CID | 169982 |
IUPAC Name | 3-chloro-4-methoxybenzoic acid |
SMILES | COC1=C(C=C(C=C1)C(=O)O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419342
|
Thermo Scientific Alfa Aesar
A10106.06 |
5 g |
£44.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429342
|
Thermo Scientific Alfa Aesar
A10106.14 |
25 g |
£153.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439342
|
Thermo Scientific Alfa Aesar
A10106.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37908-96-6 | |
186.591 | |
IBCQUQXCTOPJOD-UHFFFAOYSA-N | |
169982 | |
COC1=C(C=C(C=C1)C(=O)O)Cl |
C8H7ClO3 | |
MFCD00016512 | |
3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid | |
3-chloro-4-methoxybenzoic acid |
Specifications
37908-96-6 | |
C8H7ClO3 | |
2087584 | |
IBCQUQXCTOPJOD-UHFFFAOYSA-N | |
3-chloro-4-methoxybenzoic acid | |
169982 | |
≥98% |
215°C to 219°C | |
MFCD00016512 | |
3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid | |
COC1=C(C=C(C=C1)C(=O)O)Cl | |
186.591 | |
186.59 | |
3-Chloro-4-methoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
H301-H319
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 253-708-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only