missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methoxypyridine, 98%, Thermo Scientific Chemicals

Product Code. 15401439
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89379-79-3
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD09743650
InChI Key OFJIIJJCVDJQFW-UHFFFAOYSA-N
Synonym 4-methoxy-3-chloropyridine, 3-chloro-4-methoxy-pyridine, pyridine, 3-chloro-4-methoxy, pubchem6687
PubChem CID 23436855
IUPAC Name 3-chloro-4-methoxypyridine
SMILES COC1=C(Cl)C=NC=C1

Specifications

Quantity 5 g
Beilstein 116431
Formula Weight 143.57
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methoxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'