missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methyl-7-hydroxycoumarin, 97%, Thermo Scientific™

Product Code. 10607040
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6174-86-3
Molecular Formula C10H7ClO3
Molecular Weight (g/mol) 210.61
MDL Number MFCD00006851
InChI Key ODZHLDRQCZXQFQ-UHFFFAOYSA-N
Synonym chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl
PubChem CID 5355079
ChEBI CHEBI:38620
IUPAC Name 3-chloro-7-hydroxy-4-methylchromen-2-one
SMILES CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246.0°C to 247.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 210.62
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-chloro-4-methyl-7-hydroxycoumarin
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-methyl-7-hydroxycoumarin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'