missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Product Code. 11304666
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-41-2
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00085922
InChI Key LDSIOPGMLLPSSR-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine
PubChem CID 69996
IUPAC Name 3-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1N)Cl)[N+](=O)[O-]

Specifications

Melting Point 157°C to 161°C
Quantity 10 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 172.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'