Learn More
3-Chloro-4-nitroaniline, 95%, Thermo Scientific Chemicals
£28.50 - £164.00
Chemical Identifiers
CAS | 825-41-2 |
---|---|
Molecular Formula | C6H5ClN2O2 |
Molecular Weight (g/mol) | 172.568 |
MDL Number | MFCD00085922 |
InChI Key | LDSIOPGMLLPSSR-UHFFFAOYSA-N |
Synonym | benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine |
PubChem CID | 69996 |
IUPAC Name | 3-chloro-4-nitroaniline |
SMILES | C1=CC(=C(C=C1N)Cl)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314666
|
Thermo Scientific Alfa Aesar
H32581.03 |
1 g |
£28.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304666
|
Thermo Scientific Alfa Aesar
H32581.09 |
10 g |
£164.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
825-41-2 | |
172.568 | |
LDSIOPGMLLPSSR-UHFFFAOYSA-N | |
69996 | |
C1=CC(=C(C=C1N)Cl)[N+](=O)[O-] |
C6H5ClN2O2 | |
MFCD00085922 | |
benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine | |
3-chloro-4-nitroaniline |
Specifications
825-41-2 | |
C6H5ClN2O2 | |
UN2811 | |
Insoluble in water. | |
C1=CC(=C(C=C1N)Cl)[N+](=O)[O-] | |
172.568 | |
172.57 | |
3-Chloro-4-nitroaniline |
157°C to 161°C | |
MFCD00085922 | |
benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine | |
LDSIOPGMLLPSSR-UHFFFAOYSA-N | |
3-chloro-4-nitroaniline | |
69996 | |
95% |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only