missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluoro-4-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11481340
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-22-9
Molecular Formula C8H6ClFO3
Molecular Weight (g/mol) 204.581
MDL Number MFCD04115864
InChI Key FDWCHZNFULHBCP-UHFFFAOYSA-N
Synonym benzoic acid,3-chloro-5-fluoro-4-methoxy, 5-chloro-3-fluoro-4-methoxybenzoic acid
PubChem CID 3862547
IUPAC Name 3-chloro-5-fluoro-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1Cl)C(=O)O)F

Specifications

Quantity 1 g
Formula Weight 204.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluoro-4-methoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'