missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-fluorobenzoic acid, 97%, Thermo Scientific™
£39.58 - £221.00
Chemical Identifiers
CAS | 25026-64-6 |
---|---|
Molecular Formula | C7H4ClFO2 |
Molecular Weight (g/mol) | 174.555 |
MDL Number | MFCD00973904 |
InChI Key | QFMRANWPGGSNHS-UHFFFAOYSA-N |
Synonym | 3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 |
PubChem CID | 2734840 |
IUPAC Name | 3-chloro-5-fluorobenzoic acid |
SMILES | C1=C(C=C(C=C1F)Cl)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Chloro-5-fluorobenzoic acid is used as a pharmaceutical intermediate. It acts as a protective reagent and is used in chemical research.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
25026-64-6 | |
174.555 | |
QFMRANWPGGSNHS-UHFFFAOYSA-N | |
2734840 | |
C1=C(C=C(C=C1F)Cl)C(=O)O |
C7H4ClFO2 | |
MFCD00973904 | |
3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 | |
3-chloro-5-fluorobenzoic acid |
Specifications
25026-64-6 | |
MFCD00973904 | |
Soluble in dimethyl sulfoxide. | |
C1=C(C=C(C=C1F)Cl)C(=O)O | |
174.555 | |
174.56 | |
3-Chloro-5-fluorobenzoic acid |
C7H4ClFO2 | |
3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 | |
QFMRANWPGGSNHS-UHFFFAOYSA-N | |
3-chloro-5-fluorobenzoic acid | |
2734840 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures