Learn More
3-Chloro-5-fluorobenzoic acid, 97%, Thermo Scientific™
£35.35 - £101.00
Chemical Identifiers
CAS | 25026-64-6 |
---|---|
Molecular Formula | C7H4ClFO2 |
Molecular Weight (g/mol) | 174.56 |
MDL Number | MFCD00973904 |
InChI Key | QFMRANWPGGSNHS-UHFFFAOYSA-N |
Synonym | 3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 |
PubChem CID | 2734840 |
IUPAC Name | 3-chloro-5-fluorobenzoic acid |
SMILES | C1=C(C=C(C=C1F)Cl)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10515983
|
Thermo Scientific Acros
374760010 |
1 g | Glass bottle |
£35.35
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386412
|
Thermo Scientific Acros
374760050 |
5 g | Glass bottle | ||||||
Description
Chemical Identifiers
25026-64-6 | |
174.56 | |
QFMRANWPGGSNHS-UHFFFAOYSA-N | |
2734840 | |
C1=C(C=C(C=C1F)Cl)C(=O)O |
C7H4ClFO2 | |
MFCD00973904 | |
3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 | |
3-chloro-5-fluorobenzoic acid |
Specifications
25026-64-6 | |
100.0 | |
White to Beige | |
96% min. (GC) | |
FC6H3(Cl)COOH | |
3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247 | |
C1=C(C=C(C=C1F)Cl)C(=O)O | |
174.56 | |
174.56 | |
Crystalline Powder or Granules |
96.0 | |
153°C to 154°C | |
Authentic | |
C7H4ClFO2 | |
MFCD00973904 | |
QFMRANWPGGSNHS-UHFFFAOYSA-N | |
3-chloro-5-fluorobenzoic acid | |
2734840 | |
97% | |
3-Chloro-5-fluorobenzoic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only