Learn More
3-Chloro-5-fluorobenzonitrile, 98%, Thermo Scientific™
£19.20 - £268.00
Chemical Identifiers
CAS | 327056-73-5 |
---|---|
Molecular Formula | C7H3ClFN |
Molecular Weight (g/mol) | 155.56 |
MDL Number | MFCD03407967 |
InChI Key | GIKLRWNRBOLRDV-UHFFFAOYSA-N |
Synonym | 3-chloro-5-fluoro benzonitrile, 3-chloro-5-fluoro-benzonitrile, benzonitrile, 3-chloro-5-fluoro, 5-chloro-3-fluorobenzenecarbonitrile, 5-chloro-3-fluorobenzonitrile, pubchem4725, acmc-1afkx, 3-fluoro-5-chloro-benzonitrile, 5-fluoro-3-chloro-benzonitrile, buttpark 45\01-07 |
PubChem CID | 2734841 |
IUPAC Name | 3-chloro-5-fluorobenzonitrile |
SMILES | C1=C(C=C(C=C1F)Cl)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10462713
|
Thermo Scientific Acros
374770010 |
1 g | Glass bottle | ||||||
10598473
|
Thermo Scientific Acros
374770050 |
5 g | Glass bottle | ||||||
10075023
|
Thermo Scientific Acros
374770250 |
25 g | Glass bottle |
£268.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
327056-73-5 | |
155.56 | |
GIKLRWNRBOLRDV-UHFFFAOYSA-N | |
2734841 | |
C1=C(C=C(C=C1F)Cl)C#N |
C7H3ClFN | |
MFCD03407967 | |
3-chloro-5-fluoro benzonitrile, 3-chloro-5-fluoro-benzonitrile, benzonitrile, 3-chloro-5-fluoro, 5-chloro-3-fluorobenzenecarbonitrile, 5-chloro-3-fluorobenzonitrile, pubchem4725, acmc-1afkx, 3-fluoro-5-chloro-benzonitrile, 5-fluoro-3-chloro-benzonitrile, buttpark 45\01-07 | |
3-chloro-5-fluorobenzonitrile |
Specifications
53.0°C to 57.0°C | |
97.5 | |
100.0 | |
Crystalline Powder | |
C7H3ClFN | |
MFCD03407967 | |
GIKLRWNRBOLRDV-UHFFFAOYSA-N | |
3-chloro-5-fluorobenzonitrile | |
2734841 | |
98% |
327056-73-5 | |
Yellow | |
Authentic | |
97.5% min. (GC) | |
ClC6H3(F)CN | |
3-chloro-5-fluoro benzonitrile, 3-chloro-5-fluoro-benzonitrile, benzonitrile, 3-chloro-5-fluoro, 5-chloro-3-fluorobenzenecarbonitrile, 5-chloro-3-fluorobenzonitrile, pubchem4725, acmc-1afkx, 3-fluoro-5-chloro-benzonitrile, 5-fluoro-3-chloro-benzonitrile, buttpark 45\01-07 | |
C1=C(C=C(C=C1F)Cl)C#N | |
155.56 | |
155.56 | |
3-Chloro-5-fluorobenzonitrile |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 3-Chloro-5-fluorobenzonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only