missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. p-7029337
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11316889

Brand: Thermo Scientific Alfa Aesar H26788.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-5-(trifluoromethoxy)benzaldehyde is used as pharmaceutical intermediates

Solubility
Soluble in water (3.3 g/L (20°C)).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433926-48-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD06660305
InChI Key LWQSWEYYGGFLDR-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethoxy benzaldehyde, benzaldehyde, 3-chloro-5-trifluoromethoxy, 5-chloro-3-trifluoromethoxy benzaldehyde
PubChem CID 17750734
IUPAC Name 3-chloro-5-(trifluoromethoxy)benzaldehyde
SMILES C1=C(C=C(C=C1OC(F)(F)F)Cl)C=O

Specifications

Refractive Index 1.4765
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in water (3.3g/L (20°C)).
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.