missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11445970
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886503-33-9
Molecular Formula C8H5BrClF3O
Molecular Weight (g/mol) 289.48
MDL Number MFCD06660311
InChI Key HMVSKKRMOPJKLU-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-chloro-5-trifluoromethoxy benzene, 1-bromomethyl-3-chloro-5-trifluoromethoxy-benzene
PubChem CID 17750739
IUPAC Name 1-(bromomethyl)-3-chloro-5-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC(Cl)=CC(CBr)=C1

Specifications

Refractive Index 1.498
Quantity 1 g
UN Number UN3265
Formula Weight 289.48
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'