missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ Product Code.: 11431350

3-Chloro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11431350
10 g
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy

Product Code. 11431350

Brand: Thermo Scientific Alfa Aesar H26399.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-5-(trifluoromethyl)benzoic acid may be used as surrogate standard to investigate the exposure and home contamination in urine and hand wipe samples collected from farmers.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53985-49-2
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD04115846
InChI Key IDLOGBMWOUINGG-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl benzoic acid, 3-chloro-5-trifluoromethyl benzoicacid, 3-chloro-5-trifluoromethyl-benzoic acid, acmc-1axez, ksc495i8p, 3-carboxy-5-chlorobenzotrifluoride, 5-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 3-chloro-5-trifluoromethyl, 5-chloro-alpha,alpha,alpha-trifluoro-m-toluic acidShow MoreShow Less
PubChem CID 3802362
IUPAC Name 3-chloro-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C=C1C(F)(F)F)Cl)C(=O)O

Specifications

Melting Point 98°C to 101°C
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10145487 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10156668 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10147415 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10151177 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10141256 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.