Learn More
3-Chloro-N,N-dimethylaniline, 95%, Thermo Scientific Chemicals
£44.92 - £141.71
Chemical Identifiers
CAS | 6848-13-1 |
---|---|
Molecular Formula | C8H10ClN |
Molecular Weight (g/mol) | 155.625 |
MDL Number | MFCD00018456 |
InChI Key | CHHCCYVOJBBCIY-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride |
PubChem CID | 23285 |
IUPAC Name | 3-chloro-N,N-dimethylaniline |
SMILES | CN(C)C1=CC(=CC=C1)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352586
|
Thermo Scientific Alfa Aesar
L08563.06 |
5 g |
£44.92
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11362586
|
Thermo Scientific Alfa Aesar
L08563.14 |
25 g |
£141.71
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
6848-13-1 | |
155.625 | |
CHHCCYVOJBBCIY-UHFFFAOYSA-N | |
23285 | |
CN(C)C1=CC(=CC=C1)Cl |
C8H10ClN | |
MFCD00018456 | |
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride | |
3-chloro-N,N-dimethylaniline |
Specifications
6848-13-1 | |
C8H10ClN | |
MFCD00018456 | |
2206152 | |
Not miscible or difficult to mix in water. | |
CN(C)C1=CC(=CC=C1)Cl | |
155.625 | |
155.63 | |
3-Chloro-N,N-dimethylaniline |
230°C to 232°C | |
1.576 | |
UN2810 | |
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride | |
CHHCCYVOJBBCIY-UHFFFAOYSA-N | |
3-chloro-N,N-dimethylaniline | |
23285 | |
95% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 229-935-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 3-Chloro-N,N-dimethylaniline
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 3-Chloro-N,N-dimethylaniline
RUO – Research Use Only