missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10686354
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-04-2
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00003350
InChI Key SRWILAKSARHZPR-UHFFFAOYSA-N
Synonym m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde
PubChem CID 11477
IUPAC Name 3-chlorobenzaldehyde
SMILES C1=CC(=CC(=C1)Cl)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 17°C to 18°C
Color Colorless to Yellow
Density 1.2410g/mL
Boiling Point 213°C to 215°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CHO
Refractive Index 1.5630 to 1.565
Quantity 500 mL
Beilstein 07, 234
Specific Gravity 1.241
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 140.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-chlorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'